accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNN

CAS RN 376591-05-8

IUPAC Name: 1-benzyl-N-ethylpyrrolidin-3-amine
Synonyms: 1-benzyl-N-ethylpyrrolidin-3-amine
Molecular Weight (g/mol): 204.32
Molecular Formula: C13H20N2
InChi Key: ZSIUSRJKSLXIJH-UHFFFAOYNA-N
SMILES: CCNC1CCN(CC2=CC=CC=C2)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results