Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3R)-(-)-1-Benzyl-3-(ethylamino)pyrrolidine 95.0+%, TCI America™
SDP

Catalog No. B15805G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B15805G Supplier TCI America Supplier No. B15805G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 376591-05-8
Molecular Formula C13H20N2
Molecular Weight (g/mol) 204.317
MDL Number MFCD00191306
InChI Key ZSIUSRJKSLXIJH-CYBMUJFWSA-N
PubChem CID 12362913
IUPAC Name (3R)-1-benzyl-N-ethylpyrrolidin-3-amine
SMILES CCNC1CCN(C1)CC2=CC=CC=C2

Specifications

Color Yellow
Quantity 5 g
UN Number 2735
Formula Weight 204.32
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material (3R)-(-)-1-Benzyl-3-(ethylamino)pyrrolidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.