accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 377085-58-0

IUPAC Name: 4-carbamoyl-1-hexadecylpyridin-1-ium chloride
Synonyms: 4-carbamoyl-1-hexadecylpyridin-1-ium chloride
Molecular Weight (g/mol): 383.02
Molecular Formula: C22H39ClN2O
InChi Key: AGQLSZMFKAEZAK-UHFFFAOYSA-N
SMILES: [Cl-].CCCCCCCCCCCCCCCC[N+]1=CC=C(C=C1)C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results