accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 378-16-5

IUPAC Name: 1,1,1,2,2-pentafluoro-3-methoxypropane
Synonyms: 1,1,1,2,2-pentafluoro-3-methoxypropane
Molecular Weight (g/mol): 164.08
Molecular Formula: C4H5F5O
InChi Key: ZYAMKYAPIQPWQR-UHFFFAOYSA-N
SMILES: COCC(F)(F)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results