Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2,2,3,3,3-Pentafluoropropyl Ether 98.0+%, TCI America™
SDP

Catalog No. M25001G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25001G 1 g
M25005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M25001G Supplier TCI America Supplier No. M25001G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 378-16-5
Molecular Formula C4H5F5O
Molecular Weight (g/mol) 164.075
MDL Number MFCD00236115
InChI Key ZYAMKYAPIQPWQR-UHFFFAOYSA-N
PubChem CID 2776015
IUPAC Name 1,1,1,2,2-pentafluoro-3-methoxypropane
SMILES COCC(C(F)(F)F)(F)F

Specifications

Color Colorless
Boiling Point 48°C
Quantity 1 g
UN Number 3271
Formula Weight 164.08
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 2,2,3,3,3-Pentafluoropropyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.