accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONH2

CAS RN 37806-29-4

IUPAC Name: 1-(2-ethoxyphenyl)methanamine
Synonyms: 1-(2-ethoxyphenyl)methanamine
Molecular Weight (g/mol): 151.21
Molecular Formula: C9H13NO
InChi Key: LAUPTNYHVCVPFH-UHFFFAOYSA-N
SMILES: CCOC1=CC=CC=C1CN
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results