Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethoxybenzylamine 97.0+%, TCI America™
SDP

Catalog No. E12785G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E12785G 5 g
E12781G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E12785G Supplier TCI America Supplier No. E12785G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 37806-29-4
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.209
MDL Number MFCD00008111
InChI Key LAUPTNYHVCVPFH-UHFFFAOYSA-N
Synonym (2-Ethoxyphenyl)methanamine
PubChem CID 123465
IUPAC Name (2-ethoxyphenyl)methanamine
SMILES CCOC1=CC=CC=C1CN

Specifications

Color Yellow
Boiling Point 122°C
Quantity 5 g
UN Number 2735
Formula Weight 151.21
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Ethoxybenzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.