accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNHO

CAS RN 37812-51-4

IUPAC Name: 4-[(4-cyanophenyl)methyl]morpholin-4-ium
Synonyms: 4-[(4-cyanophenyl)methyl]morpholin-4-ium
Molecular Weight (g/mol): 203.26
Molecular Formula: C12H15N2O
InChi Key: BCZLYNFDOJXWGN-UHFFFAOYSA-O
SMILES: N#CC1=CC=C(C[NH+]2CCOCC2)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)