Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Morpholinomethyl)benzonitrile, 97%, Thermo Scientific™

Catalog No. CC29316EA
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC29316EA 10 g Amber Glass Bottle
CC29316DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC29316EA Supplier Thermo Scientific Chemicals Supplier No. CC29316EA

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 37812-51-4
Molecular Formula C12H14N2O
Molecular Weight (g/mol) 202.257
InChI Key BCZLYNFDOJXWGN-UHFFFAOYSA-N
PubChem CID 216864
IUPAC Name 4-(morpholin-4-ylmethyl)benzonitrile
SMILES C1COCCN1CC2=CC=C(C=C2)C#N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Synonym 4-morpholinomethyl benzonitrile, 4-morpholin-4-ylmethyl benzonitrile, usaf a-3730, p-tolunitrile, alpha-4-morpholinyl, alpha-4-morpholinyl-p-tolunitrile, benzonitrile, 4-4-morpholinylmethyl, 4-4-morpholinylmethyl benzonitrile, 4-morpholin-4-ylmethyl benzenecarbonitrile, 4-morpholin-4-ylmethyl-benzonitrile, benzonitrile,4-4-morpholinylmethyl
Formula Weight 202.26
Chemical Name or Material 4-(Morpholinomethyl)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.