accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONNOHNONCH3

CAS RN 37854-59-4

IUPAC Name: 7,10-dimethyl-2,4-dioxo-2H,3H,4H,10H-benzo[g]pteridine-8-carbaldehyde
Synonyms: 7,10-dimethyl-2,4-dioxo-3H-benzo[g]pteridine-8-carbaldehyde
Molecular Weight (g/mol): 270.25
Molecular Formula: C13H10N4O3
InChi Key: JDEMVNYMYPJJIM-UHFFFAOYSA-N
SMILES: CN1C2=CC(C=O)=C(C)C=C2N=C2C(=O)NC(=O)N=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)