accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOSN

CAS RN 37859-42-0

IUPAC Name: (1,3-benzothiazol-2-yl)methanol
Synonyms: 1,3-benzothiazol-2-ylmethanol
Molecular Weight (g/mol): 165.21
Molecular Formula: C8H7NOS
InChi Key: PQXMQZYDBQBWNL-UHFFFAOYSA-N
SMILES: OCC1=NC2=CC=CC=C2S1
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)