accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 37891-79-5

IUPAC Name: [bis(methylsulfanyl)methyl]trimethylsilane
Synonyms: [bis(methylsulfanyl)methyl]trimethylsilane
Molecular Weight (g/mol): 180.4
Molecular Formula: C6H16S2Si
InChi Key: QEPMPXAUMUWNNO-UHFFFAOYSA-N
SMILES: CSC(SC)[Si](C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results