accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 37968-97-1

IUPAC Name: 4-(pyridin-4-ylsulfanyl)pyridine
Synonyms: 4-(pyridin-4-ylsulfanyl)pyridine
Molecular Weight (g/mol): 188.25
Molecular Formula: C10H8N2S
InChi Key: XGJOFCCBFCHEHK-UHFFFAOYSA-N
SMILES: S(C1=CC=NC=C1)C1=CC=NC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results