accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONHOHOHNHOOHNOH

CAS RN 37988-18-4

IUPAC Name: N1,N3,N5-tris(2-hydroxyethyl)benzene-1,3,5-tricarboxamide
Synonyms: N1,N3,N5-tris(2-hydroxyethyl)benzene-1,3,5-tricarboxamide
Molecular Weight (g/mol): 339.35
Molecular Formula: C15H21N3O6
InChi Key: RGWJKANXFYJKHN-UHFFFAOYSA-N
SMILES: OCCNC(=O)C1=CC(=CC(=C1)C(=O)NCCO)C(=O)NCCO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results