accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNHNOONOONOONCH3

CAS RN 380482-30-4

IUPAC Name: 2,9-dimethyl-N-(2,4,6-trinitrophenyl)-1,10-phenanthrolin-5-amine
Synonyms: 2,9-dimethyl-N-(2,4,6-trinitrophenyl)-1,10-phenanthrolin-5-amine
Molecular Weight (g/mol): 434.37
Molecular Formula: C20H14N6O6
InChi Key: YMWCRDXLDDCILP-UHFFFAOYSA-N
SMILES: CC1=CC=C2C=C(NC3=C(C=C(C=C3[N+]([O-])=O)[N+]([O-])=O)[N+]([O-])=O)C3=CC=C(C)N=C3C2=N1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results