accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3805-67-2

IUPAC Name: 4-[2-(4-iodophenyl)diazen-1-yl]-N,N-dimethylaniline
Synonyms: 4-[2-(4-iodophenyl)diazen-1-yl]-N,N-dimethylaniline
Molecular Weight (g/mol): 351.19
Molecular Formula: C14H14IN3
InChi Key: JSCFDMNXBTVUTH-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(C=C1)N=NC1=CC=C(I)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results