accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 380560-89-4

IUPAC Name: 4-[5-(4-bromophenyl)-3-(6-methyl-2-oxo-4-phenyl-2,3-dihydroquinolin-3-ylidene)pyrazolidin-1-yl]-4-oxobutanoic acid
Synonyms: 4-[5-(4-bromophenyl)-3-(6-methyl-2-oxo-4-phenylquinolin-3-ylidene)pyrazolidin-1-yl]-4-oxobutanoic acid
Molecular Weight (g/mol): 558.43
Molecular Formula: C29H24BrN3O4
InChi Key: CADIBCWPGCNUBD-UHFFFAOYNA-N
SMILES: CC1=CC2=C(C3=CC=CC=C3)C(=C3CC(N(N3)C(=O)CCC(O)=O)C3=CC=C(Br)C=C3)C(=O)N=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results