accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 380843-75-4

IUPAC Name: 4-[(2,4-dichloro-5-methoxyphenyl)amino]-6-methoxy-7-[3-(4-methylpiperazin-1-yl)propoxy]quinoline-3-carbonitrile
Synonyms: bosutinib
Molecular Weight (g/mol): 530.45
Molecular Formula: C26H29Cl2N5O3
InChi Key: UBPYILGKFZZVDX-UHFFFAOYSA-N
SMILES: COC1=CC(NC2=C3C=C(OC)C(OCCCN4CCN(C)CC4)=CC3=NC=C2C#N)=C(Cl)C=C1Cl
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results