accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOONH

CAS RN 3814-10-6

IUPAC Name: methyl 4-(phenylcarbamoyl)benzoate
Synonyms: methyl 4-(phenylcarbamoyl)benzoate
Molecular Weight (g/mol): 255.27
Molecular Formula: C15H13NO3
InChi Key: XTAGEOVGQDJRIC-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(C=C1)C(=O)NC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results