accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 381670-32-2

IUPAC Name: 1-ethylpyrrolidine-2-carboxamide
Synonyms: 1-ethylpyrrolidine-2-carboxamide
Molecular Weight (g/mol): 142.2
Molecular Formula: C7H14N2O
InChi Key: LQOASEHNECNLNM-UHFFFAOYNA-N
SMILES: CCN1CCCC1C(N)=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results