accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 381670-34-4

IUPAC Name: 2-methylpentyl formate
Synonyms: 2-methylpentyl formate
Molecular Weight (g/mol): 130.19
Molecular Formula: C7H14O2
InChi Key: KYHHSTLLLQRFHJ-UHFFFAOYNA-N
SMILES: CCCC(C)COC=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results