accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 38172-19-9

IUPAC Name: 2-[3-(dicyanomethylidene)-2,3-dihydro-1H-inden-1-ylidene]propanedinitrile
Synonyms: 2-[3-(dicyanomethylidene)-2H-inden-1-ylidene]propanedinitrile
Molecular Weight (g/mol): 242.24
Molecular Formula: C15H6N4
InChi Key: HBZYYOYCJQHAEL-UHFFFAOYSA-N
SMILES: N#CC(C#N)=C1CC(=C(C#N)C#N)C2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results