accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CHOOFSCH3O(Z)

CAS RN 38194-50-2

IUPAC Name: 2-[(1Z)-5-fluoro-1-[(4-methanesulfinylphenyl)methylidene]-2-methyl-1H-inden-3-yl]acetic acid
Synonyms: sulindac arthrocine chemia brand of sulindac novo-sundac clinoril nu pharm brand of sulindac nu-pharm brand of sulindac kenalin sulindac sulfoxide sulindacum
Molecular Weight (g/mol): 356.41
Molecular Formula: C20H17FO3S
InChi Key: MLKXDPUZXIRXEP-MFOYZWKCNA-N
SMILES: CC1=C(CC(O)=O)C2=CC(F)=CC=C2\C1=C/C1=CC=C(C=C1)S(C)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (6)
  • (2)
Percent Purity 
  • (2)
  • (6)
Grade 
  • (1)
  • (6)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (7)
  • (1)
  • (6)
  • (2)
  • (1)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (6)
  • (2)
  • (2)
  • (6)
  • (1)
  • (6)
  • (2)
  • (1)