Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulindac, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18607181 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
10 kg
Packaging:
Amber Glass Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-181 500 g Amber Glass Bottle
18-607-176 5 g Amber Glass Bottle
18-607-177 25 g Amber Glass Bottle
18-607-178 100 g Amber Glass Bottle
18-607-179 1 kg Amber Glass Bottle
18-607-180 10 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-607-181 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S1743500GMBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

S1743, 38194-50-2, C20H17FO3S

Spectrum™ Chemical Sulindac, USP is a non-steroidal anti-inflammatory drug used to treat pain or inflammation caused by arthritis, tendinitis, bursitis, or gout. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 38194-50-2
Molecular Formula C20H17FO3S
Molecular Weight (g/mol) 356.41
InChI Key MLKXDPUZXIRXEP-MFOYZWKCNA-N
IUPAC Name 2-[(1Z)-5-fluoro-1-[(4-methanesulfinylphenyl)methylidene]-2-methyl-1H-inden-3-yl]acetic acid
SMILES CC1=C(CC(O)=O)C2=CC(F)=CC=C2\C1=C/C1=CC=C(C=C1)S(C)=O

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.005
Packaging Amber Glass Bottle
Quantity 500 g
Percent Purity 98 to 102%
Grade USP
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.