accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 382631-43-8

IUPAC Name: octane-1,2,8-triol
Synonyms: octane-1,2,8-triol
Molecular Weight (g/mol): 162.23
Molecular Formula: C8H18O3
InChi Key: GKCGJDQACNSNBB-UHFFFAOYNA-N
SMILES: OCCCCCCC(O)CO
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results