accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOOOOHHO(E/Z)

CAS RN 383177-50-2

IUPAC Name: 3-(pyridin-4-yl)prop-2-enal; oxalic acid
Synonyms: 3-(pyridin-4-yl)prop-2-enal; oxalic acid
Molecular Weight (g/mol): 223.18
Molecular Formula: C10H9NO5
InChi Key: NSTHNVMITOXEJZ-UHFFFAOYSA-N
SMILES: OC(=O)C(O)=O.O=CC=CC1=CC=NC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results