accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONHOHOH

CAS RN 383418-30-2

IUPAC Name: N-(1,4-dihydroxy-4-phenylbutan-2-yl)dodecanamide
Synonyms: N-(1,4-dihydroxy-4-phenylbutan-2-yl)dodecanamide
Molecular Weight (g/mol): 363.54
Molecular Formula: C22H37NO3
InChi Key: YCAKBKAOFSILDC-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCC(=O)NC(CO)CC(O)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results