accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHO

CAS RN 38350-87-7

IUPAC Name: 4-heptylbenzoic acid
Synonyms: 4-heptylbenzoic acid
Molecular Weight (g/mol): 220.31
Molecular Formula: C14H20O2
InChi Key: VSUKEWPHURLYTK-UHFFFAOYSA-N
SMILES: CCCCCCCC1=CC=C(C=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results