accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClONHN

CAS RN 38353-09-2

IUPAC Name: hydrogen 1,2-dihydropyrimidin-2-one chloride
Synonyms: hydrogen 2-pyrimidone chloride
Molecular Weight (g/mol): 132.55
Molecular Formula: C4H5ClN2O
InChi Key: IAJINJSFYTZPEJ-UHFFFAOYSA-N
SMILES: [H+].[Cl-].O=C1NC=CC=N1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (4)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)