accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 38361-37-4

FFCl

CAS RN 38361-37-4

IUPAC Name: 2-chloro-1,3-difluorobenzene
Synonyms: 2-chloro-1,3-difluorobenzene
Molecular Weight (g/mol): 148.54
Molecular Formula: C6H3ClF2
InChi Key: OTZQYBFTOANOJO-UHFFFAOYSA-N
SMILES: FC1=CC=CC(F)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results