CAS RN 3847-58-3
IUPAC Name:
2-chloro-1,3-difluoro-4-nitrobenzene
Synonyms:
2-chloro-1,3-difluoro-4-nitrobenzene
Molecular Weight (g/mol):
193.53
Molecular Formula:
C6H2ClF2NO2
InChi Key:
CTVBVNKEMCGZDF-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC=C(F)C(Cl)=C1F