accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 384842-25-5

IUPAC Name: di-tert-butyl(1,1-diphenylprop-1-en-2-yl)phosphane
Synonyms: di-tert-butyl(1,1-diphenylprop-1-en-2-yl)phosphane
Molecular Weight (g/mol): 338.48
Molecular Formula: C23H31P
InChi Key: CYGZMOQFIMJEIT-UHFFFAOYSA-N
SMILES: CC(P(C(C)(C)C)C(C)(C)C)=C(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)