Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

vBRIDP(regR) 98.0+%, TCI America™
SDP

Catalog No. V0132200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V0132200MG 200 mg
V01321G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V0132200MG Supplier TCI America Supplier No. V0132200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 384842-25-5
Molecular Formula C23H31P
Molecular Weight (g/mol) 338.475
MDL Number MFCD16621412
InChI Key CYGZMOQFIMJEIT-UHFFFAOYSA-N
Synonym Di-tert-butyl(1,1-diphenyl-1-propen-2-yl)phosphine
PubChem CID 12010825
IUPAC Name ditert-butyl(1,1-diphenylprop-1-en-2-yl)phosphane
SMILES CC(=C(C1=CC=CC=C1)C2=CC=CC=C2)P(C(C)(C)C)C(C)(C)C

Specifications

Melting Point 133°C
Color White-Yellow
Quantity 200 mg
Formula Weight 338.47
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material vBRIDP(regR)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.