CAS RN 3849-33-0
IUPAC Name:
1,1,1,2,3,3,3-heptachloropropane
Synonyms:
1,1,1,2,3,3,3-heptachloropropane
Molecular Weight (g/mol):
285.19
Molecular Formula:
C3HCl7
InChi Key:
LZOUTMTXULWVSU-UHFFFAOYSA-N
SMILES:
ClC(C(Cl)(Cl)Cl)C(Cl)(Cl)Cl
1
–
1
of
1
results
1
1,1,1,2,3,3,3-Heptachloropropane 98.0+%, TCI America™
CAS: 3849-33-0 Molecular Formula: C3HCl7 Molecular Weight (g/mol): 285.19 MDL Number: MFCD00058935 InChI Key: LZOUTMTXULWVSU-UHFFFAOYSA-N PubChem CID: 77457 IUPAC Name: 1,1,1,2,3,3,3-heptachloropropane SMILES: ClC(C(Cl)(Cl)Cl)C(Cl)(Cl)Cl