accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOCH3

CAS RN 3852-09-3

IUPAC Name: methyl 3-methoxypropanoate
Synonyms: methyl 3-methoxypropanoate
Molecular Weight (g/mol): 118.13
Molecular Formula: C5H10O3
InChi Key: BDJSOPWXYLFTNW-UHFFFAOYSA-N
SMILES: COCCC(=O)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results