accessibility menu, dialog, popup

UserName

Viewing profile as user:
OONS

CAS RN 38521-46-9

IUPAC Name: (3-carboxylatopyridin-2-yl)sulfanide
Synonyms: (3-carboxylatopyridin-2-yl)sulfanide
Molecular Weight (g/mol): 153.16
Molecular Formula: C6H3NO2S
InChi Key: WYKHFQKONWMWQM-UHFFFAOYSA-L
SMILES: [O-]C(=O)C1=CC=CN=C1[S-]
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)