accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 38818-50-7

IUPAC Name: 4-chloro-3-nitrobenzoyl chloride
Synonyms: 4-chloro-3-nitrobenzoyl chloride
Molecular Weight (g/mol): 220.01
Molecular Formula: C7H3Cl2NO3
InChi Key: IWLGXPWQZDOMSB-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(=CC=C1Cl)C(Cl)=O
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results