accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNH3NS

CAS RN 3882-98-2

IUPAC Name: 4,5-dihydro-1,3-thiazol-2-aminium chloride
Synonyms: 4,5-dihydro-1,3-thiazol-2-aminium chloride
Molecular Weight (g/mol): 138.61
Molecular Formula: C3H7ClN2S
InChi Key: CODJLHDDIDUKMD-UHFFFAOYSA-N
SMILES: [Cl-].[NH3+]C1=NCCS1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results