accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OCH3(S)OHHONHH2NOHO

CAS RN 38821-49-7

IUPAC Name: (2S)-3-(3,4-dihydroxyphenyl)-2-hydrazinyl-2-methylpropanoic acid hydrate
Synonyms: carbidopa hydrate (S)-(-)-carbidopa hydrate carbidopum monohydricum (S)-carbidopa hydrate
Molecular Weight (g/mol): 244.25
Molecular Formula: C10H16N2O5
InChi Key: QTAOMKOIBXZKND-PPHPATTJSA-N
SMILES: O.C[C@@](CC1=CC=C(O)C(O)=C1)(NN)C(O)=O
Molecular Weight (g/mol) 
  • (4)
  • (2)
  • (8)
Percent Purity 
  • (2)
  • (4)
  • (7)
  • (1)
Grade 
  • (1)
  • (11)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (11)
  • (4)
  • (1)
  • (7)
  • (2)
  • (1)
  • (6)
  • (4)
  • (2)
  • (8)
  • (2)
  • (4)
  • (7)
  • (1)
  • (1)
  • (11)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)