Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbidopa, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18601447 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-447 100 g Amber Glass Bottle
18-601-442 5 g Amber Glass Bottle
18-601-443 25 g Amber Glass Bottle
18-601-446 500 g Amber Glass Bottle
18-601-444 1 kg Amber Glass Bottle
18-601-445 5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-601-447 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C1401100GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

C1401, 38821-49-7, C10H14N2O4·H2O

Spectrum™ Chemical Carbidopa, USP is used in combination with levodopa to treat symptoms of Parkinson′s disease. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 38821-49-7
Molecular Formula C10H16N2O5
Molecular Weight (g/mol) 244.25
InChI Key QTAOMKOIBXZKND-PPHPATTJSA-N
IUPAC Name (2S)-3-(3,4-dihydroxyphenyl)-2-hydrazinyl-2-methylpropanoic acid hydrate
SMILES O.C[C@@](CC1=CC=C(O)C(O)=C1)(NN)C(O)=O

Specifications

CAS Max % 100%
Quantity 100 g
Ignition Residue 0.1%
Loss on Drying 6.9-7.9%
Packaging Amber Glass Bottle
Specific Rotation -21.0° to -23.5°
Percent Purity 98 to 102%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.