CAS RN 389064-25-9
CAS RN 389064-25-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ AdaAhx3L3VS
A potent, covalent, cell-permeable, irreversible inhibitor of chymotrypsin-like (50-100 nM), trypsin-like (1-5 μM), and PGPH (0.5-1 μM) activities of the 20S proteasome.