accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOOONOO

CAS RN 3899-90-9

IUPAC Name: 3-nitrophenyl 4-methylbenzene-1-sulfonate
Synonyms: 3-nitrophenyl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 293.29
Molecular Formula: C13H11NO5S
InChi Key: QRMMIQVSVATYQP-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)OC1=CC=CC(=C1)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results