CAS RN 39156-67-7
CAS RN 39156-67-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Cytochalasin B, Dihydro-
Induces changes in cell morphology and motility with no effect on sugar transport.