accessibility menu, dialog, popup

UserName

Viewing profile as user:
NFF

CAS RN 391604-55-0

IUPAC Name: 2-(2,4-difluorophenyl)pyridine
Synonyms: 2-(2,4-difluorophenyl)pyridine
Molecular Weight (g/mol): 191.18
Molecular Formula: C11H7F2N
InChi Key: SSABEFIRGJISFH-UHFFFAOYSA-N
SMILES: FC1=CC(F)=C(C=C1)C1=CC=CC=N1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)