accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOH3C

CAS RN 39212-23-2

IUPAC Name: 5-butyl-4-methyloxolan-2-one
Synonyms: 3-methyl-gamma-octalactone
Molecular Weight (g/mol): 156.23
Molecular Formula: C9H16O2
InChi Key: WNVCMFHPRIBNCW-UHFFFAOYNA-N
SMILES: CCCCC1OC(=O)CC1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results