accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 39252-69-2

IUPAC Name: 2-iodoethyl benzoate
Synonyms: 2-iodoethyl benzoate
Molecular Weight (g/mol): 276.07
Molecular Formula: C9H9IO2
InChi Key: HNQYNCREQXENCK-UHFFFAOYSA-N
SMILES: ICCOC(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results