Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Iodoethyl Benzoate, TCI America™
SDP

Catalog No. B007025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B007025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B007025ML Supplier TCI America Supplier No. B007025ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 39252-69-2
Molecular Formula C9H9IO2
Molecular Weight (g/mol) 276.073
MDL Number MFCD00059491
InChI Key HNQYNCREQXENCK-UHFFFAOYSA-N
Synonym Benzoic Acid 2-Iodoethyl Ester
PubChem CID 12510118
IUPAC Name 2-iodoethyl benzoate
SMILES C1=CC=C(C=C1)C(=O)OCCI

Specifications

Boiling Point 163°C
Quantity 25 mL
Formula Weight 276.07
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Iodoethyl Benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.