CAS RN 393-82-8
IUPAC Name:
2,5-bis(trifluoromethyl)benzoyl chloride
Synonyms:
2,5-bis(trifluoromethyl)benzoyl chloride
Molecular Weight (g/mol):
276.56
Molecular Formula:
C9H3ClF6O
InChi Key:
LRJNPOCQOZWIGR-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=CC=C(C(=C1)C(Cl)=O)C(F)(F)F
1
–
1
of
1
results
1
2,5-Bis(trifluoromethyl)benzoyl Chloride 98.0+%, TCI America™
CAS: 393-82-8 Molecular Formula: C9H3ClF6O Molecular Weight (g/mol): 276.562 MDL Number: MFCD00074997 InChI Key: LRJNPOCQOZWIGR-UHFFFAOYSA-N PubChem CID: 2737812 IUPAC Name: 2,5-bis(trifluoromethyl)benzoyl chloride SMILES: C1=CC(=C(C=C1C(F)(F)F)C(=O)Cl)C(F)(F)F