accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3HNONNNCl

CAS RN 393514-24-4

IUPAC Name: N-(4-tert-butylphenyl)-4-(3-chloropyridin-2-yl)piperazine-1-carboxamide
Synonyms: N-(4-tert-butylphenyl)-4-(3-chloropyridin-2-yl)piperazine-1-carboxamide
Molecular Weight (g/mol): 372.9
Molecular Formula: C20H25ClN4O
InChi Key: ROGUAPYLUCHQGK-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(NC(=O)N2CCN(CC2)C2=C(Cl)C=CC=N2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results