accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClCH3OONO

CAS RN 3939-01-3

IUPAC Name: hydrogen methyl 1-benzyl-4-oxopiperidine-3-carboxylate chloride
Synonyms: hydrogen methyl 1-benzyl-4-oxopiperidine-3-carboxylate chloride
Molecular Weight (g/mol): 283.75
Molecular Formula: C14H18ClNO3
InChi Key: BRADBAOVPACOQQ-UHFFFAOYNA-N
SMILES: [H+].[Cl-].COC(=O)C1CN(CC2=CC=CC=C2)CCC1=O
Molecular Weight (g/mol) 
  • (1)
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (3)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)